missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3,4-Tri-O-benzoyl-alpha-D-glucopyranoside 98.0+%, TCI America™

Catalog No. p-7132760
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14871G 1 g
M14875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14871G Supplier TCI America Supplier No. M14871G

Chemical Identifiers

CAS 34234-44-1
Molecular Formula C28H26O9
Molecular Weight (g/mol) 506.507
MDL Number MFCD06797128
InChI Key BHQKQQXWEXIQMI-NRUNVSGISA-N
PubChem CID 10918191
IUPAC Name [(2R,3R,4S,5R,6S)-4,5-dibenzoyloxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl] benzoate
SMILES COC1C(C(C(C(O1)CO)OC(=O)C2=CC=CC=C2)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
Formula Weight 506.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3,4-Tri-O-benzoyl-alpha-D-glucopyranoside
TSCA No
Recommended Storage Freezer