accessibility menu, dialog, popup

UserName

H3COOHOOOOOOO

CAS RN 34234-44-1

IUPAC Name: 4,5-bis(benzoyloxy)-2-(hydroxymethyl)-6-methoxyoxan-3-yl benzoate
Synonyms: 4,5-bis(benzoyloxy)-2-(hydroxymethyl)-6-methoxyoxan-3-yl benzoate
Molecular Weight (g/mol): 506.51
Molecular Formula: C28H26O9
InChi Key: BHQKQQXWEXIQMI-UHFFFAOYNA-N
SMILES: COC1OC(CO)C(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results