missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-dimethylacetoacetate, 99%

Catalog No. p-7045488
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1091703 1 g
AAL1091706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1091703 Supplier Thermo Scientific Chemicals Supplier No. L1091703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38923-57-8 | C7H12O3 | 144.17 g/mol

Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 38923-57-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00026232
InChI Key LVSDLZIEHYYLTC-UHFFFAOYSA-N
Synonym methyl 2,2-dimethylacetoacetate, methyl2,2-dimethylacetoacetate, butanoic acid, 2,2-dimethyl-3-oxo-, methyl ester, methyl 2,2-dimethyl acetoacetate, 2,2-dimethylacetoacetic acid methyl ester, 2,2-dimethyl-3-oxo-butyric acid methyl ester, 2,2-dimethyl-3-oxobutyric acid, methyl ester, methyl 2,2-dimethyl-3-oxobutyrate, pubchem23356, acmc-20akj7
PubChem CID 96925
IUPAC Name methyl 2,2-dimethyl-3-oxobutanoate
SMILES COC(=O)C(C)(C)C(C)=O

Specifications

Density 0.999
Boiling Point 72°C to 74°C (15 mmHg)
Refractive Index 1.4205
Quantity 1 g
Beilstein 1753940
Solubility Information Insoluble in water.
Formula Weight 144.17
Percent Purity 99%
Chemical Name or Material Methyl 2,2-dimethylacetoacetate
EINECSNumber 254-189-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only