CAS RN 38923-57-8
IUPAC Name:
methyl 2,2-dimethyl-3-oxobutanoate
Synonyms:
methyl 2,2-dimethyl-3-oxobutanoate
Molecular Weight (g/mol):
144.17
Molecular Formula:
C7H12O3
InChi Key:
LVSDLZIEHYYLTC-UHFFFAOYSA-N
SMILES:
COC(=O)C(C)(C)C(C)=O
1
–
1
of
1
results
1
Promotions Available
Methyl 2,2-dimethylacetoacetate, 99%
Methyl 2,2-dimethylacetoacetate, 99%
CAS: 38923-57-8 Molecular Formula: C7H12O3 Molecular Weight (g/mol): 144.17 MDL Number: MFCD00026232 InChI Key: LVSDLZIEHYYLTC-UHFFFAOYSA-N Synonym: methyl 2,2-dimethylacetoacetate,methyl2,2-dimethylacetoacetate,butanoic acid, 2,2-dimethyl-3-oxo-, methyl ester,methyl 2,2-dimethyl acetoacetate,2,2-dimethylacetoacetic acid methyl ester,2,2-dimethyl-3-oxo-butyric acid methyl ester,2,2-dimethyl-3-oxobutyric acid, methyl ester,methyl 2,2-dimethyl-3-oxobutyrate,pubchem23356,acmc-20akj7 PubChem CID: 96925 IUPAC Name: methyl 2,2-dimethyl-3-oxobutanoate SMILES: COC(=O)C(C)(C)C(C)=O