missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1H-indazole-6-carboxylate, 95%, Thermo Scientific™
$284.22 - $284.22
Chemical Identifiers
| CAS | 170487-40-8 |
|---|---|
| Molecular Formula | C9H8N2O2 |
| Molecular Weight (g/mol) | 176.175 |
| MDL Number | MFCD07371612 |
| InChI Key | TUSICEWIXLMXEY-UHFFFAOYSA-N |
| Synonym | 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester |
| PubChem CID | 286535 |
| IUPAC Name | methyl 1H-indazole-6-carboxylate |
| SMILES | COC(=O)C1=CC2=C(C=C1)C=NN2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3293303
|
Thermo Scientific Chemicals
H3293303 |
1 g |
Each for $284.22
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMethyl 1H-indazole-6-carboxylate is used in organic synthesis.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 170487-40-8 | |
| 176.175 | |
| TUSICEWIXLMXEY-UHFFFAOYSA-N | |
| 286535 | |
| COC(=O)C1=CC2=C(C=C1)C=NN2 |
| C9H8N2O2 | |
| MFCD07371612 | |
| 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester | |
| methyl 1H-indazole-6-carboxylate |
Specifications
| 170487-40-8 | |
| C9H8N2O2 | |
| 1 g | |
| Soluble in water. | |
| COC(=O)C1=CC2=C(C=C1)C=NN2 | |
| 176.175 | |
| 176.17 | |
| Methyl 1H-indazole-6-carboxylate |
| 142°C to 144°C | |
| MFCD07371612 | |
| 1h-indazole-6-carboxylic acid methyl ester, methyl indazole-6-carboxylate, indazole-6-carboxylic acid methyl ester, 6-1h indazole carboxylic acid methyl ester, 1h-indazole-6-carboxylic acid, methyl ester, pubchem18064, methyl indazole-6carboxylate, ksc494o4p, methyl-1h-indazole-6-carboxylate, 2h-indazole-6-carboxylic acid methyl ester | |
| TUSICEWIXLMXEY-UHFFFAOYSA-N | |
| methyl 1H-indazole-6-carboxylate | |
| 286535 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures