missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-1H-benzotriazole (mixture) 98.0+%, TCI America™
$86.64 - $531.34
Chemical Identifiers
| CAS | 29385-43-1 |
|---|---|
| Molecular Formula | C7H7N3 |
| MDL Number | MFCD00167158 |
Chemical Identifiers
| 29385-43-1 | |
| C7H7N3 | |
| MFCD00167158 |
Specifications
| 29385-43-1 | |
| White-Yellow | |
| MFCD00167158 | |
| 133.15 | |
| Crystalline Powder |
| 87°C | |
| C7H7N3 | |
| 25 g | |
| ≥98.0% (GC) | |
| Methyl-1H-benzotriazole (mixture) |
Safety and Handling
RTECSNumber : DM1300000
TSCA : No
Recommended Storage : Refrigerator