CAS RN 29385-43-1
CAS RN 29385-43-1
IUPAC Name:
4-methyl-2H-1,2,3-benzotriazole
Synonyms:
4-methyl-2H-1,2,3-benzotriazole
Molecular Weight (g/mol):
133.15
Molecular Formula:
C7H7N3
InChi Key:
CMGDVUCDZOBDNL-UHFFFAOYSA-N
SMILES:
CC1=CC=CC2=NNN=C12
1
–
1
of
1
results
1
Methyl-1H-benzotriazole (mixture) 98.0+%, TCI America™
CAS: 29385-43-1 Molecular Formula: C7H7N3 MDL Number: MFCD00167158