missing translation for 'onlineSavingsMsg'
Learn More

Methyl 10-Undecenoate 96.0+%, TCI America™

Catalog No. U003625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U003625ML 25 mL
U0036100ML 100 mL
U0036500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. U003625ML Supplier TCI America Supplier No. U003625ML

Chemical Identifiers

CAS 111-81-9
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00016689
InChI Key KISVAASFGZJBCY-UHFFFAOYSA-N
Synonym 10-Undecenoic Acid Methyl Ester
PubChem CID 8138
ChEBI CHEBI:87493
IUPAC Name methyl undec-10-enoate
SMILES COC(=O)CCCCCCCCC=C

Specifications

Color Yellow
Boiling Point 248°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 10-Undecenoate