missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylate 98.0+%, TCI America™

Catalog No. M22275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22275G 5 g
M22271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22275G Supplier TCI America Supplier No. M22275G
Only null left

Chemical Identifiers

CAS 124443-68-1
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.30
MDL Number MFCD02183584
InChI Key PTZNCCIULVXFIJ-UHFFFAOYSA-N
Synonym 1-Boc-4-piperidinecarboxylic Acid Methyl Ester, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 4-Methyl Piperidine-1,4-dicarboxylate, Methyl 1-Boc-4-piperidinecarboxylate, Piperidine-1,4-dicarboxylic Acid 1-tert-Butyl 4-Met
PubChem CID 392870
IUPAC Name 1-tert-butyl 4-methyl piperidine-1,4-dicarboxylate
SMILES COC(=O)C1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Melting Point 35°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylate