accessibility menu, dialog, popup

UserName

CAS RN 124443-68-1

CH3OONOOCH3H3CCH3

CAS RN 124443-68-1

IUPAC Name: 1-tert-butyl 4-methyl piperidine-1,4-dicarboxylate
Synonyms: 1-tert-butyl 4-methyl piperidine-1,4-dicarboxylate
Molecular Weight (g/mol): 243.3
Molecular Formula: C12H21NO4
InChi Key: PTZNCCIULVXFIJ-UHFFFAOYSA-N
SMILES: COC(=O)C1CCN(CC1)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results