missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-Methylpyrrole-2-acetate 97.0+%, TCI America™

Catalog No. M08465ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08465ML 5 mL
M084625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08465ML Supplier TCI America Supplier No. M08465ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51856-79-2
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00009752
InChI Key CGYVDYLHQYNGFC-UHFFFAOYSA-N
Synonym 1-Methylpyrrole-2-acetic Acid Methyl Ester
PubChem CID 103992
IUPAC Name methyl 2-(1-methylpyrrol-2-yl)acetate
SMILES CN1C=CC=C1CC(=O)OC

Specifications

Boiling Point 85°C
Quantity 5 mL
Formula Weight 153.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 1-Methylpyrrole-2-acetate