accessibility menu, dialog, popup

UserName

H3COONH3C

CAS RN 51856-79-2

IUPAC Name: methyl 2-(1-methyl-1H-pyrrol-2-yl)acetate
Synonyms: methyl 2-(1-methylpyrrol-2-yl)acetate
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: CGYVDYLHQYNGFC-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CN1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results