missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-cyclohexene-1-carboxylate, 97%

Catalog No. AAB2556806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2556806 5 g
AAB2556814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2556806 Supplier Thermo Scientific Chemicals Supplier No. B2556806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18448-47-0 | C8H12O2 | 140.182 g/mol

Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases. 

Chemical Identifiers

CAS 18448-47-0
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001544
InChI Key KXPWRCPEMHIZGU-UHFFFAOYSA-N
Synonym methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene
PubChem CID 87647
IUPAC Name methyl cyclohexene-1-carboxylate
SMILES COC(=O)C1=CCCCC1

Specifications

Density 1.03
Boiling Point 190°C to 192°C
Flash Point 73°C (165°F)
Refractive Index 1.477
Quantity 5 g
Solubility Information Immiscible with water.
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 1-cyclohexene-1-carboxylate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 242-331-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only