accessibility menu, dialog, popup

UserName

CAS RN 18448-47-0

IUPAC Name: methyl cyclohex-1-ene-1-carboxylate
Synonyms: methyl cyclohex-1-ene-1-carboxylate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: KXPWRCPEMHIZGU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CCCCC1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results