missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-Benzylpyrrolidine-3-carboxylate 98.0+%, TCI America™

Catalog No. M28661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28661G 1 g
M28665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28661G Supplier TCI America Supplier No. M28661G
Only null left

Chemical Identifiers

CAS 17012-21-4
Molecular Formula C13H17NO2
Molecular Weight (g/mol) 219.284
MDL Number MFCD04038678
InChI Key GDWFCUOFVSNTTG-UHFFFAOYSA-N
Synonym methyl n-benzyl-3-pyrrolidinecarboxylate, 1-benzyl-pyrrolidine-3-carboxylic acid methyl ester, methyl 1-benzyl-3-pyrrolidinecarboxylate, methyl-1-benzylpyrrolidine-3carboxylate, 3-pyrrolidinecarboxylic acid, 1-phenylmethyl-, methyl ester, methyl 1-phenylmethyl pyrrolidine-3-carboxylate, methyl n-benzylpyrrolidine-3-carboxylate, methyl 1-benzyllpyrrolidine-3-carboxylate, 1-benzylpyrrolidine-3-carboxylic acid methyl ester, 1-phenylmethyl-3-pyrrolidinecarboxylic acid methyl ester
PubChem CID 4610751
IUPAC Name methyl 1-benzylpyrrolidine-3-carboxylate
SMILES COC(=O)C1CCN(C1)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 105°C
Quantity 1 g
Formula Weight 219.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl 1-Benzylpyrrolidine-3-carboxylate