accessibility menu, dialog, popup

UserName

CH3OON

CAS RN 17012-21-4

IUPAC Name: methyl 1-benzylpyrrolidine-3-carboxylate
Synonyms: methyl 1-benzylpyrrolidine-3-carboxylate
Molecular Weight (g/mol): 219.28
Molecular Formula: C13H17NO2
InChi Key: GDWFCUOFVSNTTG-UHFFFAOYNA-N
SMILES: COC(=O)C1CCN(CC2=CC=CC=C2)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results