missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-Aminocyclopentanecarboxylate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133177
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23511G 1 g
M23515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23511G Supplier TCI America Supplier No. M23511G
Only null left

Chemical Identifiers

CAS 60421-23-0
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.64
MDL Number MFCD03840364
InChI Key OPUJUITUYWGUEP-UHFFFAOYSA-N
Synonym 1-Aminocyclopentanecarboxylic Acid Methyl Ester Hydrochloride, Cycloleucine Methyl Ester Hydrochloride
PubChem CID 11513856
IUPAC Name hydrogen methyl 1-aminocyclopentane-1-carboxylate chloride
SMILES [H+].[Cl-].COC(=O)C1(N)CCCC1

Specifications

Melting Point 203°C
Color White
Quantity 1 g
Formula Weight 179.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-Aminocyclopentanecarboxylate Hydrochloride