missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-Adamantanecarboxylate 98.0+%, TCI America™

Catalog No. M21195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21195G 5 g
M211925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21195G Supplier TCI America Supplier No. M21195G

Chemical Identifiers

CAS 711-01-3
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD01838519
InChI Key CLYOOVNORYNXMD-UHFFFAOYSA-N
Synonym 1-Adamantanecarboxylic Acid Methyl Ester
PubChem CID 136553
IUPAC Name methyl adamantane-1-carboxylate
SMILES COC(=O)C12CC3CC(C1)CC(C3)C2

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 79°C
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-Adamantanecarboxylate
TSCA No
Recommended Storage Refrigerator