accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 711-01-3

IUPAC Name: methyl adamantane-1-carboxylate
Synonyms: methyl adamantane-1-carboxylate
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: CLYOOVNORYNXMD-UHFFFAOYSA-N
SMILES: COC(=O)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results