missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,1,2,2-Tetrafluoroethyl Ether 98.0+%, TCI America™

Catalog No. M251425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M251425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M251425G Supplier TCI America Supplier No. M251425G
Only null left

Chemical Identifiers

CAS 425-88-7
Molecular Formula C3H4F4O
Molecular Weight (g/mol) 132.058
MDL Number MFCD00042106
InChI Key YQQHEHMVPLLOKE-UHFFFAOYSA-N
Synonym 1,1,2,2-tetrafluoroethyl methyl ether, methyl 1,1,2,2-tetrafluoroethyl ether, methyl1,1,2,2-tetrafluoroethylether, acmc-20ap8j, ethane, 1,1,2,2-tetrafluoro-1-methoxy, hfe-254pc, methyl 2h-tetrafluoroethyl ether, yqqhehmvplloke-uhfffaoysa, 1,1,2,2-tetrafluoroethylmethylether, 1-methoxy-1,1,2,2-tetrafluoroethane
PubChem CID 67926
IUPAC Name 1,1,2,2-tetrafluoro-1-methoxyethane
SMILES COC(C(F)F)(F)F

Specifications

Color Colorless
Boiling Point 37°C
Quantity 25 g
UN Number 3271
Formula Weight 132.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 1,1,2,2-Tetrafluoroethyl Ether
EINECSNumber (2)-3997
TSCA No