accessibility menu, dialog, popup

UserName

CAS RN 425-88-7

H3COFFFF

CAS RN 425-88-7

IUPAC Name: 1,1,2,2-tetrafluoro-1-methoxyethane
Synonyms: 1,1,2,2-tetrafluoro-1-methoxyethane
Molecular Weight (g/mol): 132.06
Molecular Formula: C3H4F4O
InChi Key: YQQHEHMVPLLOKE-UHFFFAOYSA-N
SMILES: COC(F)(F)C(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results