missing translation for 'onlineSavingsMsg'
Learn More

Methoxytrimethylsilane, 96+%

Catalog No. AC392470250
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392470250 25 g Glass bottle
AC392471000 100 g Glass bottle
AC392472500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC392470250 Supplier Thermo Scientific Chemicals Supplier No. 392470250

CAS: 1825-61-2 | C4H12OSi | 104.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1825-61-2
Molecular Formula C4H12OSi
Molecular Weight (g/mol) 104.22
MDL Number MFCD00008344,MFCD03458977
InChI Key POPACFLNWGUDSR-UHFFFAOYSA-N
Synonym trimethylmethoxysilane, silane, methoxytrimethyl, methyl trimethylsilyl ether, methoxy trimethyl silane, ch3 3sioch3, tmsome, methoxytrimethyl-silan, methoxy trimethylsilane, methoxytrimethyl silane, acmc-209ejn
PubChem CID 15771
IUPAC Name methoxy(trimethyl)silane
SMILES CO[Si](C)(C)C

Specifications

Color Colorless
Density 0.7560g/mL
Boiling Point 55.0°C to 57.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3660 to 1.3680 (20°C, 589nm)
Linear Formula H3COSi(CH3)3
Quantity 25 g
Specific Gravity 0.756
Solubility Information Solubility in water: reacts
Formula Weight 104.22
Percent Purity 96+%
Physical Form Liquid
Chemical Name or Material Methoxytrimethylsilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-369-

RUO – Research Use Only