missing translation for 'onlineSavingsMsg'
Learn More

(Methoxymethyl)triphenylphosphonium Chloride 99.0+%, TCI America™

Supplier:  TCI America M0828100G

 View more versions of this product

Catalog No. M0828100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(Methoxymethyl)triphenylphosphonium Chloride
202°C
C20H20ClOP
100 g
SJFNDMHZXCUXSA-UHFFFAOYSA-M
(methoxymethyl)triphenylphosphanium chloride
2723798
≥99.0% (HPLC)
4009-98-7
White
MFCD00011800
methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
342.80
342.80
Crystalline Powder

Chemical Identifiers

4009-98-7
342.80
SJFNDMHZXCUXSA-UHFFFAOYSA-M
2723798
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C20H20ClOP
MFCD00011800
methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride
(methoxymethyl)triphenylphosphanium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4187

TSCA : No

SDS
Documents

Documents

Promotions

Promotions