missing translation for 'onlineSavingsMsg'
Learn More

(Methoxymethyl)triphenylphosphonium Chloride 99.0+%, TCI America™

Catalog No. M0828500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0828500G 500 g
M082825G 25 g
M0828100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0828500G Supplier TCI America Supplier No. M0828500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4009-98-7
Molecular Formula C20H20ClOP
Molecular Weight (g/mol) 342.80
MDL Number MFCD00011800
InChI Key SJFNDMHZXCUXSA-UHFFFAOYSA-M
Synonym methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride
PubChem CID 2723798
IUPAC Name (methoxymethyl)triphenylphosphanium chloride
SMILES [Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 202°C
Color White
Quantity 500 g
Formula Weight 342.80
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (Methoxymethyl)triphenylphosphonium Chloride
EINECSNumber (3)-4187
TSCA No