missing translation for 'onlineSavingsMsg'
Learn More

(Methoxymethyl)triphenylphosphonium chloride, 98%

Catalog No. p-157353
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146010250 25 g Glass bottle
AC146011000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC146010250 Supplier Thermo Scientific Chemicals Supplier No. 146010250
Only null left

CAS: 4009-98-7 | C20H20ClOP | 342.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 4009-98-7
Molecular Formula C20H20ClOP
Molecular Weight (g/mol) 342.80
MDL Number MFCD00011800
InChI Key SJFNDMHZXCUXSA-UHFFFAOYSA-M
Synonym methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride
PubChem CID 2723798
IUPAC Name methoxymethyl(triphenyl)phosphanium;chloride
SMILES [Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C to 197.0°C
Color White
Boiling Point 413.0°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OCH2P(C6H5)3Cl
Quantity 25 g
Solubility Information Solubility in water: decomposes
Formula Weight 342.79
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (Methoxymethyl)triphenylphosphonium chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/f
EINECSNumber 223-664-5
TSCA TSCA

RUO – Research Use Only