missing translation for 'onlineSavingsMsg'
Learn More

(Methoxymethyl)diphenylphosphine oxide, 98+%

Catalog No. p-7043469
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0091206 5 g
AAL0091214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0091206 Supplier Thermo Scientific Chemicals Supplier No. L0091206
Only null left

CAS: 4455-77-0 | C14H15O2P | 246.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4455-77-0
Molecular Formula C14H15O2P
Molecular Weight (g/mol) 246.25
MDL Number MFCD00015451
InChI Key OEPKDBQOTLDTNC-UHFFFAOYSA-N
Synonym methoxymethyl diphenylphosphine oxide, methoxymethyldiphenylphosphine oxide, diphenyl methoxymethyl phosphine oxide, phosphine oxide, methoxymethyl diphenyl, methoxymethyl diphenyl phosphine oxide, phosphine oxide, diphenyl methoxymethyl, methoxymethyl phenyl phosphoroso benzene, acmc-1ahy3, cbmicro_008972
PubChem CID 78203
IUPAC Name [methoxymethyl(phenyl)phosphoryl]benzene
SMILES COCP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 113°C to 120°C
Quantity 5 g
Beilstein 649558
Formula Weight 246.24
Percent Purity ≥98%
Chemical Name or Material (Methoxymethyl)diphenylphosphine oxide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 224-704-4
RTECSNumber SZ0199360
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only