missing translation for 'onlineSavingsMsg'
Learn More

Methoxymethyl(diphenyl)phosphine Oxide 98.0+%, TCI America™

Catalog No. p-7132602
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12105G 5 g
M121025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12105G Supplier TCI America Supplier No. M12105G
Only null left

Chemical Identifiers

CAS 4455-77-0
Molecular Formula C14H15O2P
Molecular Weight (g/mol) 246.25
MDL Number MFCD00015451
InChI Key OEPKDBQOTLDTNC-UHFFFAOYSA-N
Synonym methoxymethyl diphenylphosphine oxide, methoxymethyldiphenylphosphine oxide, diphenyl methoxymethyl phosphine oxide, phosphine oxide, methoxymethyl diphenyl, methoxymethyl diphenyl phosphine oxide, phosphine oxide, diphenyl methoxymethyl, methoxymethyl phenyl phosphoroso benzene, acmc-1ahy3, cbmicro_008972
PubChem CID 78203
IUPAC Name [(methoxymethyl)(phenyl)phosphoroso]benzene
SMILES COCP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 115°C
Color White
Quantity 5 g
Formula Weight 246.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methoxymethyl(diphenyl)phosphine Oxide
RTECSNumber SZ0199360
TSCA No