missing translation for 'onlineSavingsMsg'
Learn More

Methoxydiphenylphosphine 98.0+%, TCI America™

Catalog No. D25575G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25575G Supplier TCI America Supplier No. D25575G

Chemical Identifiers

CAS 4020-99-9
Molecular Formula C13H13OP
Molecular Weight (g/mol) 216.22
MDL Number MFCD00048025
InChI Key OAADXJFIBNEPLY-UHFFFAOYSA-N
Synonym methoxydiphenylphosphine, methyl diphenylphosphinite, diphenylphosphinous acid methyl ester, phosphinous acid, diphenyl-, methyl ester, methoxy diphenyl phosphane, pubchem6470, acmc-209jbw, diphenyl methoxy phosphine
PubChem CID 77636
IUPAC Name methyl diphenylphosphinite
SMILES COP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 216.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxydiphenylphosphine