missing translation for 'onlineSavingsMsg'
Learn More

Methoxydimethyl(phenyl)silane 95.0+%, TCI America™

Catalog No. m29205ml
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29205ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29205ML Supplier TCI America Supplier No. M29205ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17881-88-8
Molecular Formula C9H14OSi
Molecular Weight (g/mol) 166.295
MDL Number MFCD09266142
InChI Key REQXNMOSXYEQLM-UHFFFAOYSA-N
Synonym methoxydimethylphenylsilane, methoxydimethyl phenyl silane, silane, methoxydimethylphenyl, dimethylmethoxyphenylsilane, dimethylphenylmethoxysilane, benzene, methoxydimethylsilyl, acmc-1boft, phenyldimethylmethoxysilane, amtsi001
PubChem CID 140300
IUPAC Name methoxy-dimethyl-phenylsilane
SMILES CO[Si](C)(C)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 mL
UN Number 1993
Formula Weight 166.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxydimethyl(phenyl)silane
EINECSNumber (3)-2639