accessibility menu, dialog, popup

UserName

H3COSiH3CCH3

CAS RN 17881-88-8

IUPAC Name: methoxydimethylphenylsilane
Synonyms: methoxydimethylphenylsilane
Molecular Weight (g/mol): 166.3
Molecular Formula: C9H14OSi
InChi Key: REQXNMOSXYEQLM-UHFFFAOYSA-N
SMILES: CO[Si](C)(C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results