missing translation for 'onlineSavingsMsg'
Learn More

Methoxycyclopentane (stabilized with BHT) 99.5+%, TCI America™

Catalog No. p-7132815
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1698100ML 100 mL
M1698500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1698100ML Supplier TCI America Supplier No. M1698100ML
Only null left

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point -134°C
Color Colorless
Boiling Point 106°C
Quantity 100 mL
UN Number 3271
Formula Weight 100.16
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methoxycyclopentane (stabilized with BHT)
EINECSNumber (3)-4548
TSCA Yes