missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxycarbonylsulfenyl Chloride 96.0+%, TCI America™
Supplier: TCI America M097925G
Specifications
| Methoxycarbonylsulfenyl Chloride | |
| Yellow | |
| C2H3ClO2S | |
| 25 g | |
| methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride | |
| COC(=O)SCl | |
| 126.554 | |
| 126.55 | |
| Liquid |
| 26555-40-8 | |
| 69°C | |
| MFCD00013648 | |
| 2920 | |
| TXJXPZVVSLAQOQ-UHFFFAOYSA-N | |
| methyl chlorosulfanylformate | |
| 123417 | |
| ≥96.0% (T) |
Chemical Identifiers
| 26555-40-8 | |
| 126.554 | |
| TXJXPZVVSLAQOQ-UHFFFAOYSA-N | |
| 123417 | |
| COC(=O)SCl |
| C2H3ClO2S | |
| MFCD00013648 | |
| methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride | |
| methyl chlorosulfanylformate |
Safety and Handling
TSCA : No