missing translation for 'onlineSavingsMsg'
Learn More

Methoxycarbonylsulfenyl Chloride 96.0+%, TCI America™

Catalog No. p-7132473
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09795G 5 g
M097925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09795G Supplier TCI America Supplier No. M09795G
Only null left

Chemical Identifiers

CAS 26555-40-8
Molecular Formula C2H3ClO2S
Molecular Weight (g/mol) 126.554
MDL Number MFCD00013648
InChI Key TXJXPZVVSLAQOQ-UHFFFAOYSA-N
Synonym methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride
PubChem CID 123417
IUPAC Name methyl chlorosulfanylformate
SMILES COC(=O)SCl

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 g
UN Number 2920
Formula Weight 126.55
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material Methoxycarbonylsulfenyl Chloride