missing translation for 'onlineSavingsMsg'
Learn More

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 294510050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC294510050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

29684-56-8
238.30
YSHOWEKUVWPFNR-UHFFFAOYSA-N
11032497
CC[N+](CC)(CC)S([O-])(=O)=NC(=O)OC
C8H18N2O4S
MFCD00077815,MFCD00077815,MFCD00077815
methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide
triethyl(methoxycarbonylsulfamoyl)azanium;hydroxide

Specifications

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide
Authentic
Glass bottle
CH3O2CNSO2N(C2H5)3
5g
15,1505
YSHOWEKUVWPFNR-UHFFFAOYSA-N
triethyl(methoxycarbonylsulfamoyl)azanium;hydroxide
11032497
97%
29684-56-8
96% min
C8H18N2O4S
MFCD00077815,MFCD00077815,MFCD00077815
14,208
methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide
CC[N+](CC)(CC)S([O-])(=O)=NC(=O)OC
238.30
238.31
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...