missing translation for 'onlineSavingsMsg'
Learn More

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, 97%, inner salt, Thermo Scientific™

Catalog No. p-4510463
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294510010 1g Glass bottle
AC294510050 5g Glass bottle
AC294510250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC294510010 Supplier Thermo Scientific Chemicals Supplier No. 294510010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 29684-56-8
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00077815,MFCD00077815,MFCD00077815
InChI Key YSHOWEKUVWPFNR-UHFFFAOYSA-N
Synonym methoxycarbonylsulfamoyl triethylammonium hydroxide, burgess reagent, carbomethoxysulfamoyltriethylammonium hydroxide, methoxycarbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulphamoyltriethylammonium hydroxide, methoxy-carbonylsulfamoyltriethylammonium hydroxide, methoxycarbonylsulfa-moyl-triethylammonium hydroxide, methoxycarbonylsulphamoyl-triethylammonium hydroxide
PubChem CID 11032497
IUPAC Name triethyl(methoxycarbonylsulfamoyl)azanium;hydroxide
SMILES CC[N+](CC)(CC)S([O-])(=O)=NC(=O)OC

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min
Packaging Glass bottle
Linear Formula CH3O2CNSO2N(C2H5)3
Quantity 1g
Fieser 14,208
Merck Index 15,1505
Formula Weight 238.31
Percent Purity 97%
Chemical Name or Material (Methoxycarbonylsulfamoyl)triethylammonium hydroxide
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only