missing translation for 'onlineSavingsMsg'
Learn More

(Methoxycarbonylmethyl)triphenylphosphonium bromide, 98+%

Catalog No. AAA1630136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1630136 500 g
AAA1630114 25 g
AAA1630122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1630136 Supplier Thermo Scientific Chemicals Supplier No. A1630136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1779-58-4 | C21H20BrO2P | 415.27 g/mol

(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methoxycarbonylmethyl)triphenylphosphonium bromide is a reactant for the synthesis of GSK-3 inhibitors using the Wittig reaction, substituted furanones for antifungal and cytostatic activities, photochromic dithienylethene derivatives, gem-diiodoalkanes containing allylic alcohols and highly substituted cyclopentenes and brazilanes via Au-catalyzed deoxygenative Nazarov cyclization. As reactant for asymmetric hydrogenation of α,β-unsaturated ester-phosphonates, studies of thermal decomposition of phosphonium alkyl ester salts.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 1779-58-4
Molecular Formula C21H20BrO2P
Molecular Weight (g/mol) 415.27
MDL Number MFCD00011801
InChI Key VCWBQLMDSMSVRL-UHFFFAOYSA-M
Synonym 2-methoxy-2-oxoethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenylphosphonium bromide, carbomethoxymethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenyl phosphonium bromide, 2-methoxy-2-oxoethyl triphenylphosphanium bromide, 2-methoxy-2-oxoethyl-triphenylphosphanium bromide, 2-methoxy-2-oxo-ethyl-triphenyl-phosphonium bromide, phosphonium, 2-methoxy-2-oxoethyl triphenyl-, bromide, acmc-209edx
PubChem CID 2733211
IUPAC Name (2-methoxy-2-oxoethyl)-triphenylphosphanium;bromide
SMILES [Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼163°C (decomposition)
Quantity 500 g
Beilstein 3812975
Sensitivity Hygroscopic
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 415.28
Percent Purity ≥98%
Chemical Name or Material (Methoxycarbonylmethyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-222-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only