missing translation for 'onlineSavingsMsg'
Learn More

Methoxycarbonylmethyl(triphenyl)phosphonium Bromide 97.0+%, TCI America™

Catalog No. M132625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M132625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M132625G Supplier TCI America Supplier No. M132625G

Chemical Identifiers

CAS 1779-58-4
Molecular Formula C21H20BrO2P
Molecular Weight (g/mol) 415.27
MDL Number MFCD00011801
InChI Key VCWBQLMDSMSVRL-UHFFFAOYSA-M
Synonym 2-methoxy-2-oxoethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenylphosphonium bromide, carbomethoxymethyl triphenylphosphonium bromide, methoxycarbonylmethyl triphenyl phosphonium bromide, 2-methoxy-2-oxoethyl triphenylphosphanium bromide, 2-methoxy-2-oxoethyl-triphenylphosphanium bromide, 2-methoxy-2-oxo-ethyl-triphenyl-phosphonium bromide, phosphonium, 2-methoxy-2-oxoethyl triphenyl-, bromide, acmc-209edx
PubChem CID 2733211
IUPAC Name (2-methoxy-2-oxoethyl)triphenylphosphanium bromide
SMILES [Br-].COC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 415.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methoxycarbonylmethyl(triphenyl)phosphonium Bromide