missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate

Catalog No. p-7042595
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63075MC 100 mg
AAJ6307503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63075MC Supplier Thermo Scientific Chemicals Supplier No. J63075MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-05-2 | C20H22N8O5 | 454.447 g/mol

Methotrexate is used as s cell cycle arresting agent that inhibits DHFR. It is used in treatment of cancer, autoimmune diseases, ectopic pregnancy, and for the induction of medical abortions. It acts by inhibiting the metabolism of folic acid. Methotrexate is one of the most effective and widely used medications for treating rheumatoid arthritis (RA) and other inflammatory types of arthritis. It is used as an anticancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute HCl or dilute alkali hydroxide solutions (i.e. 0.1M NaOH). Insoluble in plain water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents. Protect from light

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
MDL Number MFCD00150847
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Color Yellow
Quantity 100 mg
UN Number UN1544
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,5985
Solubility Information Soluble in dilute HCl or dilute alkali hydroxide solutions (i;e; 0;1M NaOH); Insoluble in plain water.
Formula Weight 454.44
Physical Form Powder
Chemical Name or Material Methotrexate
Hazard Category H301-H315-H319-H335-H340-H360FD
Hazard Statement GHS H Statement
H301-H360-H315-H319
Toxic if swallowed.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber 200-413-8
RTECSNumber MA1225000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only