missing translation for 'onlineSavingsMsg'
Learn More

Méthotrexate hydrate 98,0+%, TCI America™

Numéro de catalogue. M16641G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M16641G 1 g
M16645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M16641G Fournisseur TCI America Code fournisseur M16641G

Identifiants chimiques

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
MDL Number MFCD00150847
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminoptéridin-6-yl)méthyl-méthylamino]benzoyl]amino]pentanedioïque
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Spécifications

Melting Point 205°C
Quantity 1 g
UN Number 2811
Formula Weight 454.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material Methotrexate Hydrate
RTECSNumber MA1225000
TSCA Yes