missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Methionol | |
| Yellow | |
| C4H10OS | |
| 25 g | |
| CZUGFKJYCPYHHV-UHFFFAOYSA-N | |
| 3-(methylsulfanyl)propan-1-ol | |
| 10448 | |
| 106.18 | |
| Liquid |
| 505-10-2 | |
| 136°C | |
| MFCD00036560 | |
| methionol, 3-methylthiopropanol, 3-methylthio-1-propanol, 3-hydroxypropyl methyl sulfide, 3-methylthio propanol, 3-methylmercapto-1-propanol, 1-propanol, 3-methylthio, 3-methylsulfanyl propan-1-ol, gamma-methylmercaptopropyl alcohol | |
| CSCCCO | |
| 106.18 | |
| CHEBI:49019 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 505-10-2 | |
| 106.18 | |
| CZUGFKJYCPYHHV-UHFFFAOYSA-N | |
| 10448 | |
| 3-(methylsulfanyl)propan-1-ol |
| C4H10OS | |
| MFCD00036560 | |
| methionol, 3-methylthiopropanol, 3-methylthio-1-propanol, 3-hydroxypropyl methyl sulfide, 3-methylthio propanol, 3-methylmercapto-1-propanol, 1-propanol, 3-methylthio, 3-methylsulfanyl propan-1-ol, gamma-methylmercaptopropyl alcohol | |
| CHEBI:49019 | |
| CSCCCO |
Safety and Handling
EINECSNumber : (2)-0469
TSCA : Yes