missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Methimazole USP
Description
Methimazole USP is a white to pale buff, crystalline powder which has a faint characteristic odor.
Specifications
Specifications
| Chemical Name or Material | Methimazole USP |
| Packaging | Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT |
| CAS | 60-56-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 114.17 |
| Quantity | 500 g |
| Percent Purity | 98 to 101 % |
| Grade | USP |