missing translation for 'onlineSavingsMsg'
Learn More

Methimazole, ≥99%, MilliporeSigma™ Supelco™

Catalog No. p-7187464
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012225 25 g
111012226 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012225 Supplier Merck Emd Millipore Supplier No. M850625G
Only null left

Methimazole is an antithyroid agent, widely used in the treatment of hyperthyroidism.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 60-56-0
Physical Form Neat
Linear Formula C4H6N2 S
MDL Number MFCD00179321
Melting Point 144°C to 147°C
Synonym 1-Methyl-2-imidazolethiol; 2-Mercapto-1-methylimidazole
Type Analytical Standard
Quantity 25 g
Formula Weight 114.17
Percent Purity ≥99%
Grade Analytical Standard
Show More Show Less