missing translation for 'onlineSavingsMsg'
Learn More

Methanol/Water Mobile Phase 85% HPLC Methanol, Reagents

Catalog No. 01334713
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334713 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334713 Supplier Labchem Inc Supplier No. CM1279004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methanol/Water Mobile Phase 85% HPLC Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 66°C
Percent Purity 85%
Grade Reagent
Chemical Name or Material Methanol/Water Mobile Phase 85% HPLC Methanol