missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Synthetic (Methyl Alcohol), Reagents
Methanol, Synthetic (Methyl Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C212100020E6
Specifications
| Methanol, Synthetic (Methyl Alcohol) | |
| 67-56-1 | |
| 64.5°C | |
| Steel Pail | |
| 0.7866 | |
| Methyl Alcohol, Wood Alcohol | |
| CO | |
| 32.04 |
| -98°C | |
| Colorless | |
| 20 L | |
| Liquid | |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| Reagent |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| Methyl Alcohol, Wood Alcohol | |
| CO |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |