missing translation for 'onlineSavingsMsg'
Learn More

Methanol/Reagent Alcohol, 95/5 v/v, Reagents

Catalog No. p-200021771
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334715 1 L
01334714 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334715 Supplier Labchem Inc Supplier No. CM1277001A
Only null left

Methanol/Reagent Alcohol, 95/5 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 64-17-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.7931
Grade Reagent
Chemical Name or Material Methanol/Reagent Alcohol, 95/5 v/v