missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, HR-GC grade, 99.9% min., OmniSolv, MilliporeSigma™
Supplier: Technidata MX04801
Specifications
| Methanol | |
| 67-56-1 | |
| Liquid | |
| CH4O | |
| MFCD00004595 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.4 |
| OmniSolv™ HR-GC | |
| 4 L | |
| 99.9%, min. | |
| CH3OH | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| 32.04 | |
| CHEBI:17790 |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |
Safety and Handling
Recommended Storage : Room Temperature