missing translation for 'onlineSavingsMsg'
Learn More

Methanol, HR-GC grade, 99.9% min., OmniSolv, MilliporeSigma™

Catalog No. p-4382218
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX04806 1 L
MMX04801 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMX04806 Supplier Technidata Supplier No. MX04806
Only null left

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Quantity 1 L
Physical Form Liquid
Assay Percent Range 99.9%, min.
Linear Formula CH3OH
Formula Weight 32.4
Chemical Name or Material Methanol, OmniSolv™ HR-GC
Recommended Storage Room Temperature