missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents
Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1121000200E
Specifications
| Methanol, ACS Reagent, Absolute (Methyl Alcohol, Anhydrous) | |
| 67-56-1 | |
| 64.5°C | |
| Steel Drum | |
| 0.7866 | |
| Methyl Alcohol, Wood Alcohol | |
| CO | |
| 32.04 |
| -98°C | |
| Colorless | |
| 200 L | |
| Liquid | |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| ACS |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| Methyl Alcohol, Wood Alcohol | |
| CO |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |