missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents

Catalog No. p-200021203
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
Poly Bottle
Steel Drum
Steel Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338141 10 L Steel Pail
01338140 1 L Glass Bottle
01338139 2.5 L Glass Bottle
01338138 200 L Steel Drum
01338137 20 L Steel Pail
01338136 4 L Poly Bottle
01338135 4 L Glass Bottle
01338134 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01338141 Supplier Labchem Inc Supplier No. C112100010E6
Only null left

Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Boiling Point 64.5°C
Quantity 10 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.7866
Grade ACS
Chemical Name or Material Methanol, ACS Reagent, Absolute (Methyl Alcohol, Anhydrous)