missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents
Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1121000500C
Specifications
| Methanol, ACS Reagent, Absolute (Methyl Alcohol, Anhydrous) | |
| 67-56-1 | |
| 64.5°C | |
| Glass Bottle | |
| 0.7866 | |
| Methyl Alcohol, Wood Alcohol | |
| CO | |
| 32.04 |
| -98°C | |
| Colorless | |
| 500 mL | |
| Liquid | |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| ACS |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| Methyl Alcohol, Wood Alcohol | |
| CO |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |